Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002944.2 | NP_002935.2 | 28 - 29 | 4872 | 98 | |
XM_006715548.3 | XP_006715611.1 | 29 - 30 | 4964 | 98 | ||
XM_011536049.2 | XP_011534351.1 | 29 - 30 | 5010 | 98 | ||
XM_011536050.2 | XP_011534352.1 | 29 - 30 | 5007 | 98 | ||
XM_011536051.2 | XP_011534353.1 | 28 - 29 | 4983 | 98 | ||
XM_011536052.2 | XP_011534354.1 | 29 - 30 | 4968 | 98 | ||
XM_011536053.2 | XP_011534355.1 | 29 - 30 | 4836 | 98 | ||
XM_011536054.2 | XP_011534356.1 | 29 - 30 | 5011 | 98 | ||
XM_011536055.2 | XP_011534357.1 | 29 - 30 | 5010 | 98 | ||
XM_011536056.2 | XP_011534358.1 | 29 - 30 | 5009 | 98 | ||
XM_017011172.1 | XP_016866661.1 | 28 - 29 | 4941 | 98 | ||
XM_017011173.1 | XP_016866662.1 | 28 - 29 | 5040 | 98 | ||
GenBank mRNA | M34353.1 | - | 28 - 29 | 4879 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Homo sapiens Probe spans exons |