Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
TC05001891.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
ENST00000274496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
ENST00000377986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
ENST00000448443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
ENST00000513112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
ENST00000536767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026127.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012354.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000274496.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000448443.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000508754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000513112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000519064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
ENST00000522203.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
NM_001024947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
NM_001271732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
NM_030799.8 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500185938.hg.1 |
TC0500012354.hg.1 |
Clariom D TC |