Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009150.hg.1 |
ENST00000284061.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009150.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009150.hg.1 |
TC17000699.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000570738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000570738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000570738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162291.hg.1 |
TC1700008357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000284061.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000570738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000570738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000570738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000571084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000572944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
ENST00000576869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008357.hg.1 |
NM_003647.2 |
Refseq |