Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015884.hg.1 |
TC06001327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015884.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015884.hg.1 |
ENST00000378198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015884.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015884.hg.1 |
ENST00000545780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015884.hg.1 |
ENST00000545995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015884.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015882.hg.1 |
TC06001327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015882.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015882.hg.1 |
ENST00000378198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015882.hg.1 |
ENST00000545780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015882.hg.1 |
ENST00000545995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015882.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174889.hg.1 |
TC0600011067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174891.hg.1 |
TC0600011067.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000341060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000378198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000478507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
ENST00000480495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011067.hg.1 |
NM_016614.2 |
Refseq |