Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031869.hg.1 |
TC03001757.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031869.hg.1 |
ENST00000296255.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031869.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031869.hg.1 |
ENST00000497289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031869.hg.1 |
ENST00000537139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031869.hg.1 |
ENST00000545956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031869.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000479113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000479113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000479113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192360.hg.1 |
TC0300012346.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000296255.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000476931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000479113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000479113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000479113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000481168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000490166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000495462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
ENST00000497415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012346.hg.1 |
NM_002950.3 |
Refseq |