Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005084.hg.1 |
TC01000333.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005084.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151847.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151847.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151847.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151847.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151847.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151847.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151847.hg.1 |
TC0100007439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000374271.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000444713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
ENST00000619836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243532.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_001243533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007439.hg.1 |
NM_152835.4 |
Refseq |