Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003182.2 | NP_003173.1 | 5 - 6 | 534 | 68 | |
GenBank mRNA | AF050655.1 | - | 4 - 5 | 183 | 68 | |
AK222957.1 | - | 5 - 6 | 409 | 68 | ||
BC018047.1 | - | 5 - 6 | 408 | 4792746 | 68 | |
CR407602.1 | - | 4 - 5 | 288 | 68 | ||
CR541730.1 | - | 4 - 5 | 288 | 68 | ||
DN992454.1 | - | 5 - 6 | 542 | 68 | ||
U37529.1 | - | 5 - 6 | 433 | 68 | ||
X54469.1 | - | 5 - 6 | 369 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |