Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
ENST00000373656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_001304364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011454.hg.1 |
NM_020924.3 |
Refseq |