Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025684.hg.1 |
TC07003369.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025684.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025684.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025681.hg.1 |
TC07003369.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025681.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025681.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025683.hg.1 |
TC07003369.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025683.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025683.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201328.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201328.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201328.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201328.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201328.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201328.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201328.hg.1 |
TC0700013595.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700201330.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201330.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201330.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201330.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201330.hg.1 |
TC0700013595.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
ENST00000418952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013595.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201331.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201331.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201331.hg.1 |
ENST00000332375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201331.hg.1 |
NM_001004323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201331.hg.1 |
TC0700013595.hg.1 |
Clariom D TC |