Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178858.4 | NP_849189.1 | 5 - 6 | 679 | 74 | |
XM_011539261.2 | XP_011537563.1 | 6 - 7 | 680 | 74 | ||
XM_011539262.2 | XP_011537564.1 | 6 - 7 | 800 | 74 | ||
XM_011539263.2 | XP_011537565.1 | 7 - 8 | 844 | 74 | ||
XM_011539264.2 | XP_011537566.1 | 6 - 7 | 706 | 74 | ||
GenBank mRNA | AF462052.1 | - | 5 - 6 | 634 | 74 | |
AK308246.1 | - | 5 - 6 | 584 | 74 | ||
BC022091.2 | - | 5 - 6 | 679 | 3029622 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |