Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005255317.4 | XP_005255374.1 | 1 - 2 | 76 | 84 | |
XM_017023231.1 | XP_016878720.1 | 1 - 2 | 86 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |