Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000536.hg.1 |
TC0Y000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000536.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000536.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000535.hg.1 |
TC0Y000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000535.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000535.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144979.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144979.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144979.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144979.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144979.hg.1 |
TC0Y00006581.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006581.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144980.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144980.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144980.hg.1 |
ENST00000445253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144980.hg.1 |
NR_001539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144980.hg.1 |
TC0Y00006581.hg.1 |
Clariom D TC |