Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030974.hg.1 |
TC01002003.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030974.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030974.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190310.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190310.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190310.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190310.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190310.hg.1 |
TC0100012278.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
ENST00000366510.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012278.hg.1 |
NM_016002.2 |
Refseq |