Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146039.1 | NP_001139511.1 | 2 - 3 | 519 | 69 | |
NM_001320252.1 | NP_001307181.1 | 4 - 5 | 819 | 69 | ||
NM_152281.2 | NP_689494.2 | 2 - 3 | 519 | 69 | ||
XM_011510149.2 | XP_011508451.1 | 2 - 3 | 456 | 69 | ||
GenBank mRNA | AK021814.1 | - | 2 - 3 | 512 | 69 | |
BC047476.1 | - | 4 - 5 | 790 | 5296051 | 69 | |
BC064945.1 | - | 2 - 3 | 510 | 6019822 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |