Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001790.hg.1 |
ENST00000025301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001790.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001790.hg.1 |
TC13000152.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147471.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147471.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147471.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147471.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147471.hg.1 |
TC1300006989.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
ENST00000025301.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006989.hg.1 |
NM_016248.3 |
Refseq |