Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233193.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233193.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233193.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233193.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233193.hg.1 |
TC0100017625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233192.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233192.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233192.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233192.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233192.hg.1 |
TC0100017625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233194.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233194.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233194.hg.1 |
TC0100017625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233195.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233195.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233195.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233195.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233195.hg.1 |
TC0100017625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
ENST00000595971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017625.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233191.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233191.hg.1 |
NM_001287262.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233191.hg.1 |
TC0100017625.hg.1 |
Clariom D TC |
Human Genome U95 Set |
84989_at |
- |
- |
Human Genome U95 Set |
84989_PM_at |
- |
- |