Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006405.hg.1 |
TC09000575.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006405.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006405.hg.1 |
ENST00000374165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006405.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
ENST00000490544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008490.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
ENST00000288462.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
ENST00000374165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
NM_001278629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
NM_001278630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
NM_152786.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156701.hg.1 |
TC0900008490.hg.1 |
Clariom D TC |