Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028373.hg.1 |
TC06002256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028373.hg.1 |
ENST00000367144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028373.hg.1 |
ENST00000400788.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028373.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028371.hg.1 |
TC06002256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028371.hg.1 |
ENST00000367144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028371.hg.1 |
ENST00000400788.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028371.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028372.hg.1 |
TC06002256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028372.hg.1 |
ENST00000367144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028372.hg.1 |
ENST00000400788.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028372.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028369.hg.1 |
TC06002256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028369.hg.1 |
ENST00000367144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028369.hg.1 |
ENST00000400788.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028369.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028370.hg.1 |
TC06002256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028370.hg.1 |
ENST00000367144.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028370.hg.1 |
ENST00000400788.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028370.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028374.hg.1 |
TC06002256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028374.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007974.hg.1 |
ENST00000546650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194433.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194433.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194433.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194433.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194433.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194433.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194433.hg.1 |
TC0600013682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600194432.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194432.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194432.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194432.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194432.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194432.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194432.hg.1 |
TC0600013682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600194436.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194436.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194436.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194436.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194436.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194436.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194436.hg.1 |
TC0600013682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600194437.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194437.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194437.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194437.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194437.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194437.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194437.hg.1 |
TC0600013682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600194438.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194438.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194438.hg.1 |
TC0600013682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600194430.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194430.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194430.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194430.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194430.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194430.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194430.hg.1 |
TC0600013682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007974.hg.1 |
ENST00000546650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013682.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194434.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194434.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194434.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194434.hg.1 |
ENST00000367144.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194434.hg.1 |
ENST00000400788.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194434.hg.1 |
NM_018452.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194434.hg.1 |
TC0600013682.hg.1 |
Clariom D TC |