Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001815.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001815.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001815.hg.1 |
TC11000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001817.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001817.hg.1 |
ENST00000533426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001817.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001817.hg.1 |
TC11000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001816.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001816.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001816.hg.1 |
TC11000128.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100146907.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146907.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146907.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146907.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146907.hg.1 |
TC1100006721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006721.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146905.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146905.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146905.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146905.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146905.hg.1 |
TC1100006721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100146908.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146908.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146908.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146908.hg.1 |
ENST00000379936.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146908.hg.1 |
ENST00000533426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146908.hg.1 |
NM_001037329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146908.hg.1 |
TC1100006721.hg.1 |
Clariom D TC |