Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000247295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000372271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
ENST00000464831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011769.hg.1 |
NM_033387.3 |
Refseq |