Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141919.1 | NP_001135391.1 | 2 - 3 | 325 | 125 | |
NM_001141920.1 | NP_001135392.1 | 2 - 3 | 325 | 125 | ||
NM_175569.2 | NP_780778.1 | 2 - 3 | 325 | 125 | ||
XM_005274587.3 | XP_005274644.1 | 2 - 3 | 1253 | 125 | ||
XM_011545575.2 | XP_011543877.1 | 2 - 3 | 1269 | 125 | ||
GenBank mRNA | AF380356.1 | - | 2 - 3 | 303 | 125 | |
AF534880.1 | - | 2 - 3 | 299 | 125 | ||
BC100764.2 | - | 3 - 4 | 290 | 228 | ||
BC100765.2 | - | 2 - 3 | 187 | 40000158 | 125 | |
BC100766.2 | - | 2 - 3 | 188 | 40000159 | 125 | |
BC100767.1 | - | 2 - 3 | 187 | 40000160 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |