Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRDV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK301114.1 | - | 132 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens 1700 |