Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014354.hg.1 |
ENST00000340356.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014354.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014354.hg.1 |
TC20001045.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014355.hg.1 |
ENST00000340356.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014355.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014355.hg.1 |
TC20001045.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172036.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172036.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172036.hg.1 |
TC2000010035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000172036.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172036.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172041.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172041.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172041.hg.1 |
TC2000010035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000172041.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172041.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172039.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172039.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172039.hg.1 |
TC2000010035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000172039.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172039.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172040.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172040.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172040.hg.1 |
TC2000010035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000172040.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172040.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010035.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172037.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172037.hg.1 |
NM_018419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000172037.hg.1 |
TC2000010035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000172037.hg.1 |
ENST00000340356.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000172037.hg.1 |
NM_018419.2 |
Refseq |