Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022266.hg.1 |
ENST00000317578.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022266.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022266.hg.1 |
TC19001631.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022263.hg.1 |
ENST00000317578.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022263.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022263.hg.1 |
TC19001631.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022264.hg.1 |
ENST00000317578.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022264.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022264.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022264.hg.1 |
TC19001631.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022267.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022267.hg.1 |
TC19001631.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022265.hg.1 |
ENST00000317578.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022265.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022265.hg.1 |
TC19001631.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900191087.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191087.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191087.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191087.hg.1 |
TC1900010953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191087.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191087.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191087.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191092.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191092.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191092.hg.1 |
TC1900010953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191092.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191092.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191085.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191085.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191085.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191085.hg.1 |
TC1900010953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191085.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191085.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191085.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191086.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191086.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191086.hg.1 |
TC1900010953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191086.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191086.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
TC1900010953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191084.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191091.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191091.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191091.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191091.hg.1 |
TC1900010953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191091.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191091.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191091.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191083.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191083.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191083.hg.1 |
NM_175875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191083.hg.1 |
TC1900010953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191083.hg.1 |
ENST00000317578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191083.hg.1 |
ENST00000560168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191083.hg.1 |
NM_175875.4 |
Refseq |