Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033655.3 | NP_387504.2 | 10 - 11 | 1889 | 122 | |
GenBank mRNA | AB051501.2 | - | 10 - 11 | 1796 | 122 | |
AF333769.2 | - | 10 - 11 | 1757 | 122 | ||
AK056833.1 | - | 10 - 11 | 1889 | 122 | ||
BC132737.1 | - | 10 - 11 | 1794 | 40146759 | 122 |
Gene Symbol: CNTNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201380.2 | NP_001188309.2 | 10 - 11 | 2060 | 122 |
Gene Symbol: CNTNAP3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111893.2 | 10 - 11 | 1749 | 122 |
Gene Symbol: LOC101930090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017015388.1 | XP_016870877.1 | 1 - 2 | 234 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |