Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000686.4 | NP_000677.2 | 3 | 2372 | 135 | |
XM_011537533.1 | XP_011535835.1 | 2 | 2270 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |