Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000275364.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000407904.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000447791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000447791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000447791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000471281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000471281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000471281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000471281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000471281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000471281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000471281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000485329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000485329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000485329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000491117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
ENST00000496740.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001282441.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_001293092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010075.hg.1 |
NM_007353.2 |
Refseq |