Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR9LP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026850.1 | 5 - 6 | 1876 | 67 | ||
GenBank mRNA | AK096109.1 | - | 4 - 5 | 1428 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | eukaryotic translation initiation factor 3 subunit I | EIF3I |
EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36 |
-438.0 | Anti-sense | Chr. 1 - 32221584 - 32231604 on NCBI Build GRCh38 |
Downstream | family with sequence similarity 167 member B | FAM167B | C1orf90 | 12.4kb | Anti-sense | Chr. 1 - 32247217 - 32248860 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |