Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AATK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027255.1 | 2 - 3 | 407 | 84 | ||
GenBank mRNA | AK126811.1 | - | 2 - 3 | 407 | 84 | |
BC140737.1 | - | 2 - 3 | 398 | 84 | ||
BC171777.1 | - | 2 - 3 | 398 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | apoptosis associated tyrosine kinase | AATK |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
-605.0 | Anti-sense | Chr. 17 - 81117295 - 81166072 on NCBI Build GRCh38 |
Downstream | centrosomal protein 131 | CEP131 |
AZ1, AZI1, ZA1 |
10.4kb | Anti-sense | Chr. 17 - 81189593 - 81222989 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |