Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GDP-D-glucose phosphorylase 1 | GDPGP1 |
C15orf58, VTC2 |
-3.3kb | Sense | Chr. 15 - 90234255 - 90242080 on NCBI Build GRCh38 |
Downstream |
tubulin tyrosine ligase like 13, pseudogene |
TTLL13P | TTLL13 | 3.7kb | Sense | Chr. 15 - 90249532 - 90259088 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |