Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SPECC1L-ADORA2A readthrough (NMD candidate) | SPECC1L-ADORA2A | Anti-sense | Chr. 22 - 24270817 - 24442360 on NCBI Build GRCh38 | ||
Downstream | beta-ureidopropionase 1 | UPB1 | BUP1 | 54.2kb | Anti-sense | Chr. 22 - 24495060 - 24528681 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |