Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | forkhead box K1 | FOXK1 | FOXK1L | Sense | Chr. 7 - 4644691 - 4771443 on NCBI Build GRCh38 | |
Downstream | adaptor related protein complex 5 zeta 1 subunit | AP5Z1 |
KIAA0415, SPG48, zeta |
15.1kb | Sense | Chr. 7 - 4775617 - 4794395 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |