Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153790.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153790.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153790.hg.1 |
TC2100007980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153790.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153790.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153791.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153791.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153791.hg.1 |
TC2100007980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153791.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153791.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007980.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153792.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153792.hg.1 |
NR_109961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153792.hg.1 |
TC2100007980.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153792.hg.1 |
ENST00000421051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153792.hg.1 |
NR_109961.1 |
Refseq |