Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methyltransferase like 16 | METTL16 | METT10D | -100.5kb | Anti-sense | Chr. 17 - 2416049 - 2511906 on NCBI Build GRCh38 |
Downstream | clustered mitochondria homolog | CLUH |
CLU1, KIAA0664 |
76.3kb | Anti-sense | Chr. 17 - 2689386 - 2711633 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |