Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | thioredoxin reductase 3 neighbor | TXNRD3NB |
TR2IT1, TXNRD3IT1, TXNRD3NT1 |
-15.9kb | Sense | Chr. 3 - 126571779 - 126608555 on NCBI Build GRCh38 |
Downstream | nucleoporin 210 pseudogene 1 | NUP210P1 | C3orf46 | 34.8kb | Anti-sense | Chr. 3 - 126662081 - 126671939 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |