Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SR-related CTD associated factor 8 | SCAF8 | RBM16 | -5.9kb | Sense | Chr. 6 - 154733378 - 154834244 on NCBI Build GRCh38 |
Downstream | microRNA 1273c | MIR1273C | 11.2kb | Sense | Chr. 6 - 154853360 - 154853436 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |