Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000483520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000483520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
ENST00000483520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000334937.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000338637.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376950.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000376954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000483520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000483520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
ENST00000483520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007158.hg.1 |
NM_007137.3 |
Refseq |