Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIMS binding protein 3C | RIMBP3C |
RIM-BP3.3, RIMBP3.3 |
-63.7kb | Anti-sense | Chr. 22 - 21545666 - 21551461 on NCBI Build GRCh38 |
Downstream | YdjC homolog (bacterial) | YDJC | 12.1kb | Anti-sense | Chr. 22 - 21628089 - 21630326 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |