Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105373065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC043011.1 | - | 1 | 143 | 96 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BQ068032.1 | - | 155 | Chr. - - [-] on Build | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KIAA0930 | KIAA0930 |
C22orf9, LSC3 |
-21.5kb | Sense | Chr. 22 - 45192242 - 45240769 on NCBI Build GRCh38 |
Downstream | uroplakin 3A | UPK3A |
UP3A, UPIII, UPIIIA, UPK3 |
21.4kb | Anti-sense | Chr. 22 - 45284987 - 45295874 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |