Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012329.hg.1 |
ENST00000360358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012329.hg.1 |
NM_001001957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031087.hg.1 |
TC01002016.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031087.hg.1 |
ENST00000360358.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031087.hg.1 |
ENST00000537741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01031087.hg.1 |
NM_001001957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007389.hg.1 |
ENST00000406496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190571.hg.1 |
TC0100012329.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012329.hg.1 |
ENST00000360358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012329.hg.1 |
NM_001001957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007389.hg.1 |
ENST00000406496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190572.hg.1 |
ENST00000360358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190572.hg.1 |
NM_001001957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190572.hg.1 |
ENST00000360358.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100190572.hg.1 |
NM_001001957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100190572.hg.1 |
TC0100012329.hg.1 |
Clariom D TC |