Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007740.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007740.hg.1 |
ENST00000545448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007740.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007740.hg.1 |
TC10000675.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000158001.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158001.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158001.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158001.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158001.hg.1 |
TC1000008508.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000260682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000461906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
ENST00000473496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008508.hg.1 |
NM_000771.3 |
Refseq |