Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019392.hg.1 |
ENST00000334221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019392.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019392.hg.1 |
TC12001477.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180811.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180811.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180811.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180811.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180811.hg.1 |
TC1200010626.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010626.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180812.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180812.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180812.hg.1 |
ENST00000334221.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180812.hg.1 |
NM_001008223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180812.hg.1 |
TC1200010626.hg.1 |
Clariom D TC |