Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010733.hg.1 |
TC09000823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010733.hg.1 |
NR_036251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009237.hg.1 |
ENST00000585012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009237.hg.1 |
NR_036251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163043.hg.1 |
ENST00000585012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163043.hg.1 |
NR_036251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163043.hg.1 |
ENST00000585012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163043.hg.1 |
NR_036251.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163043.hg.1 |
TC0900009237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009237.hg.1 |
ENST00000585012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009237.hg.1 |
NR_036251.1 |
Refseq |