Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301009.1 | NP_001287938.1 | 1 - 2 | 486 | 102 | |
NM_001317843.1 | NP_001304772.1 | 1 - 2 | 486 | 102 | ||
NM_182546.3 | NP_872352.2 | 1 - 2 | 486 | 102 | ||
XM_006715665.3 | XP_006715728.1 | 1 - 2 | 747 | 102 | ||
XM_006715666.3 | XP_006715729.1 | 1 - 2 | 747 | 102 | ||
XM_017011841.1 | XP_016867330.1 | 1 - 2 | 747 | 102 | ||
GenBank mRNA | BC028404.1 | - | 1 - 2 | 361 | 4820705 | 102 |
BI755592.1 | - | 1 - 2 | 486 | 102 | ||
DN991717.1 | - | 1 - 2 | 248 | 102 | ||
DN991972.1 | - | 1 - 2 | 486 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |