Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000267853.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000380565.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000460962.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000461709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000468886.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000471563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000484300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000488175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000496627.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NM_001018100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NM_152451.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NR_104367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NR_104371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000267853.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000380565.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000460962.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000461709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000468886.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000471563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000484300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000488175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
ENST00000496627.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NM_001018100.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NM_152451.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NR_104367.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
NR_104371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182483.hg.1 |
TC1500010744.hg.1 |
Clariom D TC |