Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000149632.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000149632.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000149632.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000149632.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000149632.hg.1 |
TC1000007282.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007282.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000149630.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000149630.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000149630.hg.1 |
ENST00000626667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000149630.hg.1 |
NR_110138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000149630.hg.1 |
TC1000007282.hg.1 |
Clariom D TC |