Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AX-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046592.2 | 1 - 2 | 269 | 125 | ||
GenBank mRNA | AI221979.1 | - | 1 - 2 | 125 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | small Cajal body-specific RNA 9-like | SCARNA9L | -3.6kb | Anti-sense | Chr. X - 20136066 - 20136413 on NCBI Build GRCh38 | |
Downstream | ribosomal protein S6 kinase A3 | RPS6KA3 |
CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2 |
9.5kb | Anti-sense | Chr. X - 20149911 - 20267514 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |