Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541940.2 | XP_005541997.2 | 4 - 5 | 871 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |