Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015447715.1 | XP_015303201.1 | 2 - 3 | 890 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |