Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102128656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566724.2 | XP_005566781.1 | 3 - 4 | 1687 | 67 | |
XM_015456855.1 | XP_015312341.1 | 4 - 5 | 1973 | 67 | ||
GenBank mRNA | AB070021.1 | - | 4 - 5 | 1176 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |